2,5-Dithiobiurea 97%
100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 142-46-1 Molecular Formula: C2H6N4S2 Formula Weight: 150.22 Melting Point: ca 212° dec. Beilstein Reference: 1723178 MDL No.: MFCD00014471
Synonyms: Hydrazine-1,2-bis(carbothioamide)
- CAS Number:142-46-1
- Formula:C₂H₆N₄S₂
- MDL Number:MFCD00014471
- Molecular Weight:150.22 g/mol
- Melting Pt:∼ 212 °C (dec.)
- Storage Temperature:Ambient
- EINECS:205-537-6
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pk:100 g



