Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
3,5-Dihydroxybenzaldehyde ≥98%

3,5-Dihydroxybenzaldehyde ≥98%

Glass bottle for solids, 5 g
Catalog # A15338.06
Irritant
3,5-Dihydroxybenzaldehyde ≥98%
Catalog # A15338.06
CAS Number:  26153-38-8
3,5-Dihydroxybenzaldehyde ≥98%
3,5-Dihydroxybenzaldehyde ≥98%

Glass bottle for solids, 5 g

CAS Number:  26153-38-8
Price
£195.00
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98% CAS No.: 26153-38-8 Molecular Formula: C7H6O3 Formula Weight: 138.12 Physical Form: Powder Sensitivity: Air Sensitive Beilstein Reference: 1930147 MDL No.: MFCD00016611

  • CAS Number:26153-38-8
  • Formula:C₇H₆O₃
  • MDL Number:MFCD00016611
  • Molecular Weight:138.12 g/mol
  • Melting Pt:158…161 °C
  • Storage Temperature:Ambient
  • EINECS:247-479-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.