2-Fluorophenol ≥98%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 367-12-4 Molecular Formula: C6H5FO Formula Weight: 112.10 Flash Point: 46°(114°F) Refractive Index: 1.5140 Odour: Phenol-like Melting Point: 14-16° Boiling Point: 150-152° Beilstein Reference: 1905112 UN No.: 2924 MDL No.: MFCD00002155
Synonyms: o-Fluorophenol
- CAS Number:367-12-4
- Formula:C₆H₅FO
- MDL Number:MFCD00002155
- Molecular Weight:112.1 g/mol
- Boiling Pt:150…152 °C
- Melting Pt:14…16 °C
- Density:1,215
- Flash Pt:46 °C (114 °F)
- Storage Temperature:Ambient
- EINECS:206-681-2
- UN:2924
- ADR:3,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



