β-Cyclodextrine hydrate
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
β-Cyclodextrine hydrate
Catalog # A14529.22
Supplier: Thermo Fisher Scientific
CAS Number: 68168-23-0
Jump to:
Product Details & Documents
CAS No.: 68168-23-0 Molecular Formula: C42H70O35.xH2O Formula Weight: 1134.98(anhy) Melting Point: 288° dec. Beilstein Reference: 5915513 Merck Reference: 14,2718 Optical Rotation: +142° (c=1.5 in water) MDL No.: MFCD00150811
- CAS Number:68168-23-0
- Formula:C₄₂H₇₀O₃₅·H₂O
- MDL Number:MFCD00150811
- Molecular Weight:1134.98 g/mol (anhy)
- Merck Index:14,2718
- Melting Pt:288 °C (dec.)
- Storage Temperature:Ambient
- EINECS:231-493-2
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



