β-Cyclodextrine hydrate
Glass bottle for solids, 5 g
CAS No.: 68168-23-0 Molecular Formula: C42H70O35.xH2O Formula Weight: 1134.98(anhy) Melting Point: 288° dec. Beilstein Reference: 5915513 Merck Reference: 14,2718 Optical Rotation: +142° (c=1.5 in water) MDL No.: MFCD00150811
- :68168-23-0
- :C₄₂H₇₀O₃₅·H₂O
- :MFCD00150811
- :1134.98 g/mol (anhy)
- :14,2718
- :288 °C (dec.)
- :Ambient
- :231-493-2
- Pack type:Glass bottle for solids
- Pk:5 g