Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
2'-Nitroacetophenone ≥97%

2'-Nitroacetophenone ≥97%

Glass bottle for solids, 50 g
Catalog # A13208.18
Irritant
2'-Nitroacetophenone ≥97%
Catalog # A13208.18
CAS Number:  577-59-3
2'-Nitroacetophenone ≥97%
2'-Nitroacetophenone ≥97%

Glass bottle for solids, 50 g

CAS Number:  577-59-3
Price
£186.00
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 97% CAS No.: 577-59-3 Molecular Formula: C8H7NO3 Formula Weight: 165.15 Flash Point: 110°(230°F) Refractive Index: 1.5515 Odour: Characteristic Melting Point: 25-28° Boiling Point: 158-160°/16mm Beilstein Reference: 1102322 MDL No.: MFCD00007144

Synonyms:  o-Nitroacetophenone, 1-(2-Nitrophenyl)ethanone
  • CAS Number:577-59-3
  • Formula:C₈H₇NO₃
  • MDL Number:MFCD00007144
  • Molecular Weight:165.15 g/mol
  • Boiling Pt:158…160 °C (16 mmHg)
  • Melting Pt:25…28 °C
  • Density:1,24
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:209-414-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.