2'-Nitroacetophenone ≥97%
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
2'-Nitroacetophenone ≥97%
Catalog # A13208.09
Supplier: Thermo Fisher Scientific
CAS Number: 577-59-3
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 577-59-3 Molecular Formula: C8H7NO3 Formula Weight: 165.15 Flash Point: 110°(230°F) Refractive Index: 1.5515 Odour: Characteristic Melting Point: 25-28° Boiling Point: 158-160°/16mm Beilstein Reference: 1102322 MDL No.: MFCD00007144
Synonyms: o-Nitroacetophenone, 1-(2-Nitrophenyl)ethanone
- CAS Number:577-59-3
- Formula:C₈H₇NO₃
- MDL Number:MFCD00007144
- Molecular Weight:165.15 g/mol
- Boiling Pt:158…160 °C (16 mmHg)
- Melting Pt:25…28 °C
- Density:1,24
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:209-414-8
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Cat. No.:A13208.09



