ContactUsLinkComponent
Flask volumetric, capacity 200ml, class A, amber graduated, polystoppered, tolerance 0.15ml, height 210mm, NS-ground 14/23, graduati on color white, conformity certified,DIN/ISO product with single certificate. 1 * 1 items
Catalog # HIRS264018508
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 200ml, class A, amber graduated, polystoppered, tolerance 0.15ml, height 210mm, NS-ground 14/23, graduati on color white, conformity certified,DIN/ISO product with single certificate. 1 * 1 items
Catalog # HIRS264018508
Supplier:  HIRSCHMANN
Frequently Bought Together