ContactUsLinkComponent
Flask volumetric, capacity 5ml, class A, amber graduated, glass stoppered, tolerance 0.025ml, height 70mm, NS-ground 7/16, graduatio n color white, conformity certified. 1 * 1 items
Catalog # HIRS2640252_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 5ml, class A, amber graduated, glass stoppered, tolerance 0.025ml, height 70mm, NS-ground 7/16, graduatio n color white, conformity certified. 1 * 1 items
Catalog # HIRS2640252_S
Supplier:  HIRSCHMANN
Frequently Bought Together