Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Microplate 96 well polystyrene black with µClear F-bottom streptavidin-coated non-sterile 1 * 40 items
Catalog # 392-0033
Supplier:
Greiner Bio-One
Expand image
Microplate 96 well polystyrene black with µClear F-bottom streptavidin-coated non-sterile 1 * 40 items
Catalog # 392-0033
Supplier:
Greiner Bio-One
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR®, Conical Sample Containers, with Snap-On Lids
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection