ContactUsLinkComponent
Laboratory pH meter Lab 850. Measuring parameters pH, mV, temp., microprocessor, DIN 19262 connection. Including cover Z 880, stand S4D Z 865 and power supply Z 850. 1 * 1 items
Catalog # 663-0083
Supplier:  SI Analytics
Laboratory pH meter Lab 850. Measuring parameters pH, mV, temp., microprocessor, DIN 19262 connection. Including cover Z 880, stand S4D Z 865 and power supply Z 850. 1 * 1 items
Catalog # 663-0083
Supplier:  SI Analytics
Frequently Bought Together