Fused silica cutting tools
Carborundum glass file
VARICP22672
: VARIAN
This high quality, long life tungsten carbide glass knife produces perfect cuts, and is especially suitable for use with CP-Quick-Seal connectors.
- Description:Carborundum glass file
- Seq:03