ContactUsLinkComponent
2,2'-azobis-2-methyl-propanimidamide, dihydrochlorid AAPH is a water-soluble azo compound which is used extensively as a free radical generator, often in the study of lipid peroxidation and the characterization of antioxidants. 1 * 10 g
Catalog # CAYM82235-10
Supplier:  Cayman Chemical
2,2'-azobis-2-methyl-propanimidamide, dihydrochlorid AAPH is a water-soluble azo compound which is used extensively as a free radical generator, often in the study of lipid peroxidation and the characterization of antioxidants. 1 * 10 g
Catalog # CAYM82235-10
Supplier:  Cayman Chemical
Frequently Bought Together