Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Histosette® I, M491

Biopsy processing/embedding cassettes, Histosette® I, M491

Turquoise
Catalog # 720-0897
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Histosette® I, M491
Catalog # 720-0897
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Histosette® I, M491
Biopsy processing/embedding cassettes, Histosette® I, M491

Turquoise

Price
£241.00
Log in for availability
1 * 1.500 items
Product Details & Documents

Acetal polymer.

  • Resistant to all commonly used histological solvents
  • Square openings (1 mm) to maximise fluid exchange and ensure proper drainage
  • Integral lid
  • Anterior writing area at a 30° angle
Specifications
  • Colour:
    Turquoise
  • Seq:
    12
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.