ContactUsLinkComponent
Vials Short Thread 0.2ml 32x11.6mm TPX with integrated glass micro insert pack1000 1 * 1.000 items
Catalog # LAPH11141189
Supplier:  LA PHA PACK
Vials Short Thread 0.2ml 32x11.6mm TPX with integrated glass micro insert pack1000 1 * 1.000 items
Catalog # LAPH11141189
Supplier:  LA PHA PACK
Frequently Bought Together