ISOVERSINIC TUBING 12/17 ISO 1 * 1 m
Catalog # GERH10-0091
Supplier: GERHARDT
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Accessories for Mettler balances
Supplier: Mettler - Toledo
VWR® SP100 Automatic Straw Pipette, Fixed Volume
Supplier: VWR Collection
Accessories for ThermoMixer™ F1,5 and FP
Supplier: EPPENDORF
Titration beakers
Supplier: Mettler - Toledo
VWR®, Hand Tally Counter
Supplier: VWR Collection
Accessories for cell counter, EVE™
Supplier: NANOENTEK
Silicone antifoam, Supelco®
Supplier: Merck
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
pH electrodes, SenTix® PLUS
Supplier: WTW
VWR® Facial Tissues
Supplier: VWR Collection
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
Grade 113 qualitative filter papers, Whatman™
Supplier: Whatman products (Cytiva)
Titrators, SIMPLEX
Supplier: GERBER FUNKE
Plastic cuvette for LICO
Supplier: Hach