Orders LinkContactUsLinkComponent
PCR microtubes with flat cap, Axygen®
PCR microtubes with flat cap, Axygen®
PCR microtubes with flat cap, 0,2 ml, Green
Catalog # 731-4001
Supplier:   Corning
PCR microtubes with flat cap, Axygen®
Catalog # 731-4001
Supplier:  Corning
PCR microtubes with flat cap, Axygen®
PCR microtubes with flat cap, Axygen®
PCR microtubes with flat cap, 0,2 ml, Green
Price
Inquire For Price
Log in for availability
1 * 1.000 items
Product Details & Documents
Polypropylene tubes are specifically designed for PCR and thermal transfer applications.
  • Made from highly polished molds
  • Free of lubricants, dyes, heavy metals, and fillers
  • Certified RNase-, DNase-, and pyrogen-free to ensure the purity of your reaction
The ultra-thin and consistent tube walls ensure that thermal transfer is precisely even along the surface of the tube. The unique cap design guarantees a perfect fit to prevent sample evaporation during thermal cycling.
Specifications
  • Colour:
    Green
  • Description:
    PCR microtubes with flat cap, 0,2 ml
  • Seq:
    02