CERAMIC VALVE 4-2 FOR PSD4 1 * 1 items
Catalog # HAMI9234-01
Supplier: HAMILTON BONADUZ
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Filter protection 5 ml 1 * 50 items
Supplier: EPPENDORF
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Accessories for PIPETBOY acu / pro
Supplier: Integra
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
Silicone sealing rings with PTFE washer DURAN®
Supplier: DWK Life Sciences
1000 and 1700 series Gastight® syringes for PSD/4 syringe pumps
Supplier: HAMILTON BONADUZ