Areometer, for milk of lime, measuring range:0-340g/l, without thermometer, length:290mm. 1 * 1 items
Catalog # AMARH860688
Supplier: Amarell
Areometer, for milk of lime, measuring range:0-340g/l, without thermometer, length:290mm. 1 * 1 items
Catalog # AMARH860688
Supplier: Amarell
Frequently Bought Together
VWR® Measuring Cylinders, Low Form, Class B, Borosilicate Glass
Supplier: VWR Collection
Volumetric flasks, USP, class A, blue markings, BLAUBRAND®
Supplier: Brand
VWR®, Hydrometers, M100
Supplier: VWR Collection
Volumetric flasks, class A, white markings, BLAUBRAND®
Supplier: Brand
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
Measuring cylinders, tall form, class A, borosilicate glass
Supplier: HIRSCHMANN
VWR® Graduated Pipettes, Type 3, Class AS
Supplier: VWR Collection
VWR®, Burettes, Straight Stopcock
Supplier: VWR Collection
Bulb pipettes (one mark), BLAUBRAND®, class AS
Supplier: Brand
VWR®, Bulb Pipettes, One Mark, Class AS
Supplier: VWR Collection
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Supplier: VWR Collection