ContactUsLinkComponent
Thermometer, digital, measuring range: -50:150/0.1C, robust piercing probe of stainless steel with tapered top 125x3.5/5mm, maximum /minimum function, water resistance, automatic power off after 10 minutes. 1 * 1 items
Catalog # AMARE905230
Supplier:  Amarell
Thermometer, digital, measuring range: -50:150/0.1C, robust piercing probe of stainless steel with tapered top 125x3.5/5mm, maximum /minimum function, water resistance, automatic power off after 10 minutes. 1 * 1 items
Catalog # AMARE905230
Supplier:  Amarell
Frequently Bought Together