γ-Cyclodextrine hydrate
Glass bottle for solids, 100 mg
Supplier: Thermo Fisher Scientific
γ-Cyclodextrine hydrate
Catalog # L11271.MC
Supplier: Thermo Fisher Scientific
CAS Number: 91464-90-3
Jump to:
Product Details & Documents
CAS No.: 91464-90-3 Molecular Formula: C48H80O40.xH2O Formula Weight: 1297.15(anhy) Melting Point: ca 267° dec. Beilstein Reference: 5725162 Merck Reference: 14,2718 Optical Rotation: +177° (c=1 in water) on dry basis MDL No.: MFCD00149574
Synonyms: γ-Cyclodextrin hydrate, γ-Cyclodextrin xhydrate
- CAS Number:91464-90-3
- Formula:C₄₈H₈₀O₄₀·H₂O
- MDL Number:MFCD00149574
- Molecular Weight:1297.15 g/mol (anhy)
- Merck Index:14,2718
- Melting Pt:∼ 267 °C (dec.)
- Storage Temperature:Ambient
- EINECS:241-482-4
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



