ContactUsLinkComponent
LABCOLD Freezer, Sparkfree, Front Opening, 45 Litre Capacity, 525mm (h) x 500mm (w) x 500mm (d), Optional Door Lock and Temperature Alarm 1 * 1 items
Catalog # 472-1031
Supplier:  Labcold
LABCOLD Freezer, Sparkfree, Front Opening, 45 Litre Capacity, 525mm (h) x 500mm (w) x 500mm (d), Optional Door Lock and Temperature Alarm 1 * 1 items
Catalog # 472-1031
Supplier:  Labcold
Frequently Bought Together