Orders LinkContactUsLinkComponent
Separating funnels, pear shaped
Separating funnels, pear shaped
500 ml, 24/29 NS
Catalog # 221-0065
Supplier:   JONES FR
Separating funnels, pear shaped
Catalog # 221-0065
Supplier:  JONES FR
Separating funnels, pear shaped
Separating funnels, pear shaped
500 ml, 24/29 NS
Price
£98.30
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass, with glass stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.
Specifications
  • Socket size:
    24/29 NS
  • Capacity:
    500 ml