Orders LinkContactUsLinkComponent
Separating funnels, pear shaped
Separating funnels, pear shaped
250 ml, 19/26 NS
Catalog # 221-0064
Supplier:   JONES FR
Separating funnels, pear shaped
Catalog # 221-0064
Supplier:  JONES FR
Separating funnels, pear shaped
Separating funnels, pear shaped
250 ml, 19/26 NS
Price
£91.90
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass, with glass stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    250 ml