Orders LinkContactUsLinkComponent
Separating funnels, pear shaped
50 ml, 19/26 NS
  221-0060
 :   JONES FR
Separating funnels, pear shaped
  221-0060
 :  JONES FR
Separating funnels, pear shaped
Separating funnels, pear shaped
50 ml, 19/26 NS
£61.40
1 * 1 items
Borosilicate glass, with glass stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.
  • Socket size:
    19/26 NS
  • Capacity:
    50 ml