Permanent marker, Sharpie
Permanent marker, fine point, Blue
Catalog # 811-0024
Supplier: DIALSTAT OFFICE SUPPLIES
Jump to:
Product Details & Documents
These Sharpie markers with long lasting durable fibre tip are permanent on most hard to mark surfaces.
- Fine tip line width of 1 mm for precision marking
- Fade and water resistant
- Quick-drying ink
Excellent for marking cardboard/photo paper/corrugate/wood/metal and foil.
Specifications
- Colour:Blue
- Description:Permanent marker, fine point