Non return valves, Azlon®
11-15 mm ID
Catalog # AZLONWH906P
Supplier: SciLabware
Jump to:
Product Details & Documents
Designed for use in vacuum lines and fluid handling systems.
- Useful for preventing back pressure in filter flasks
- Printed arrow indicates the direction of flow
- Rigid with stepped tapered arms to give firm grip on tubing
Specifications
- Tubing size:11-15 mm ID