DILTIAZEM HYDROCHLORIDE IMP. E (EP): 1 * 100 mg
Catalog # PROOMM0068.04
Supplier: LGC Standards PROMOCHEM
DILTIAZEM HYDROCHLORIDE IMP. E (EP): 1 * 100 mg
Catalog # PROOMM0068.04
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
n-Hexane, EMPLURA®, Supelco®
Supplier: Merck
Ethanol absolute, SupraSolv® for GC-ECD/FID
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Ethanol 96% (v/v), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
(±)-2-Butanol, EMPLURA®, Supelco®
Supplier: Merck
Methanol, EMPLURA®, Supelco®
Supplier: Merck
Methanol ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
Supplier: VWR Chemicals
Methanol ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
1-Pentanol 99%, pure
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP), extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclopentyl methyl ether (CPME) ≥99.9%
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals