Orders LinkContactUsLinkComponent
Forceps
Forceps
Straight, fine tips, 115 mm
Catalog # 232-0034
Supplier:   MURRAY S
Forceps
Catalog # 232-0034
Supplier:  MURRAY S
Forceps
Forceps
Straight, fine tips, 115 mm
Price
£7.10
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel.
  • Sharp, narrow flutes
  • With guide pin
Specifications
  • Type:
    Straight, fine tips
  • Length:
    115 mm