Orders LinkContactUsLinkComponent
Replacement impeller for Celstir® flasks
Impeller Retrofit for 500 ml flask
  WHEA356903
Replacement impeller for Celstir® flasks
  WHEA356903
Replacement impeller for Celstir® flasks
Replacement impeller for Celstir® flasks
Impeller Retrofit for 500 ml flask
£246.00
1 * 1 items
This unit includes a top cap with liners, glass rod, magnet holder, magnet and stirring paddle (for sizes 125 ml or larger).
  • Description:
    Impeller Retrofit for 500 ml flask
  • Seq:
    06