Orders LinkContactUsLinkComponent
Replacement impeller for Celstir® flasks
Impeller for 125 ml flask
  734-3161
Replacement impeller for Celstir® flasks
  734-3161
Replacement impeller for Celstir® flasks
Replacement impeller for Celstir® flasks
Impeller for 125 ml flask
£264.00
1 * 1 items
This unit includes a top cap with liners, glass rod, magnet holder, magnet and stirring paddle (for sizes 125 ml or larger).
  • Description:
    Impeller for 125 ml flask
  • Seq:
    03