Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Replacement impeller for Celstir® flasks

Replacement impeller for Celstir® flasks

Impeller for 500 ml flask
Catalog # 734-3163
Supplier:   DWK Life Sciences
Replacement impeller for Celstir® flasks
Catalog # 734-3163
Supplier:  DWK Life Sciences
Replacement impeller for Celstir® flasks
Replacement impeller for Celstir® flasks

Impeller for 500 ml flask

Price
£325.00
Log in for availability
1 * 1 items
Product Details & Documents

This unit includes a top cap with liners, glass rod, magnet holder, magnet and stirring paddle (for sizes 125 ml or larger).

Specifications
  • Description:
    Impeller for 500 ml flask
  • Seq:
    05