Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Container, with lid

Container, with lid

5000 ml, 285 mm, 175 mm
Catalog # BOCH8081
Supplier:   BOCHEM
Container, with lid
Catalog # BOCH8081
Supplier:  BOCHEM
Container, with lid
Container, with lid

5000 ml, 285 mm, 175 mm

Price
£928.00
Log in for availability
1 * 1 items
Product Details & Documents

Stainless steel 18/10

  • Double walled
  • Stackable
  • With handle
Specifications
  • Ø opening:
    175 mm
  • Capacity:
    5000 ml
  • Height:
    285 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.