Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Double bossheads

Double bossheads

Double bosshead, safety screw type 'heart' M8, Aluminium, 16,5 mm
Catalog # BOCH5356
Supplier:   BOCHEM
Double bossheads
Catalog # BOCH5356
Supplier:  BOCHEM
Double bossheads
Double bossheads

Double bosshead, safety screw type 'heart' M8, Aluminium, 16,5 mm

Price
£23.50
Log in for availability
1 * 1 items
Product Details & Documents

Double bossheads, either made of stainless steel or aluminium.

  • Angle: 180 + 90°
  • Different screw types available
Specifications
  • Description:
    Double bosshead, safety screw type 'heart' M8
  • Material:
    Aluminium
  • For rod Ø:
    16,5 mm