Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
n-Hexane ≥95%, TECHNICAL

n-Hexane ≥95%, TECHNICAL

Metal can, 5 L
Catalog # 24611.366
Flammable
Health
Toxic
Environment
Supplier:   VWR Chemicals
n-Hexane ≥95%, TECHNICAL
Catalog # 24611.366
Supplier:  VWR Chemicals
CAS Number:  110-54-3
n-Hexane ≥95%, TECHNICAL
n-Hexane ≥95%, TECHNICAL

Metal can, 5 L

CAS Number:  110-54-3
Price
£110.00
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:13,04712
  • Boiling Pt:69 °C (1013 hPa)
  • Melting Pt:–94,3 °C
  • Density:0,659 g/cm³ (20 °C)
  • Flash Pt:–22 °C
  • Storage Temperature:Ambient
  • EINECS:203-777-6
  • UN:1208
  • ADR:3,II
  • REACH:01-2119480412-44
Specifications

Specification Test Results

  • Assay:
    Min. 95 %
  • Boiling point:
    68 - 70 °C
  • Benzene:
    Max. 100 ppm
  • Evaporation residue:
    Max. 20 ppm

Specifications

  • Pack type:
    Metal can
  • Pk:
    5 L

Specifications