n-Hexane ≥95%, TECHNICAL
Glass bottle, 1 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Hexyl hydride, Hexanen, Hexane
- CAS Number:110-54-3
- Formula:H₃C(CH₂)₄CH₃
- MDL Number:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Boiling Pt:69 °C (1013 hPa)
- Melting Pt:–94,3 °C
- Density:0,659 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Storage Temperature:Ambient
- EINECS:203-777-6
- UN:1208
- ADR:3,II
- REACH:01-2119480412-44
Documents
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 5L Tin bottle for liquids UN Y (ZPACK12321007)
Packaging Information
Specifications
Specification Test Results
- Assay:Min. 95 %
- Boiling point:68 - 70 °C
- Benzene:Max. 100 ppm
- Evaporation residue:Max. 20 ppm
Specifications
- Pack type:Glass bottle
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



