Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
(±)-1,2-Propanediol ≥99%, TECHNICAL

(±)-1,2-Propanediol ≥99%, TECHNICAL

Plastic bottle, 1 L
Catalog # 24414.296
Supplier:   VWR Chemicals
(±)-1,2-Propanediol ≥99%, TECHNICAL
Catalog # 24414.296
Supplier:  VWR Chemicals
CAS Number:  57-55-6
(±)-1,2-Propanediol ≥99%, TECHNICAL
(±)-1,2-Propanediol ≥99%, TECHNICAL

Plastic bottle, 1 L

CAS Number:  57-55-6
Price
£45.60
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
  • CAS Number:57-55-6
  • Formula:CH₃CH(OH)CH₂OH
  • MDL Number:MFCD00064272
  • Molecular Weight:76.1 g/mol
  • Merck Index:13,07947
  • Boiling Pt:187,6 °C (1013 hPa)
  • Melting Pt:–59 °C
  • Density:1,04 g/cm³ (20 °C)
  • Flash Pt:99 °C
  • Storage Temperature:Ambient
  • EINECS:200-338-0
Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Refractive index (20°C):
    1.431 - 1.435

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.