Wipe rolls, Multiclean®
Roll with embossing, 2 ply, Blue, 360×220 mm, 2 rolls of 1000 sheets (380 m per roll)
Catalog # 115-2735
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Jump to:
Product Details & Documents
These perforated paper towel rolls are suitable for the removal of major dirt on surfaces. They are suitable for industry and trade applications, e.g. machine maintenance.
- High absorbency and wet strength
- 2 or 3 ply
- Can be used with floor-standing and wall-mountable dispensers
Colour: Blue.
Specifications
- Packed:2 rolls of 1000 sheets (380 m per roll)
- Colour:Blue
- Description:Roll with embossing, 2 ply
- L×W:360×220 mm
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



