ContactUsLinkComponent
Thermometer, maximum, with index, mercury filled, measuring range:-10:250/1C, length:300mm, diameter:9-10mm, colourless, capillary lens front, with ring at top, not allowed to immerse into temperature bath upto the reading. 1 * 1 items
Catalog # 620-1513
Supplier:  Amarell
Thermometer, maximum, with index, mercury filled, measuring range:-10:250/1C, length:300mm, diameter:9-10mm, colourless, capillary lens front, with ring at top, not allowed to immerse into temperature bath upto the reading. 1 * 1 items
Catalog # 620-1513
Supplier:  Amarell
Frequently Bought Together