Filter paper folded MN 611 1/4 18.5cm diameter, pack of 100. 1 * 100 items
Catalog # 516-5010
Supplier: MACHEREY-NAGEL
Filter paper folded MN 611 1/4 18.5cm diameter, pack of 100. 1 * 100 items
Catalog # 516-5010
Supplier: MACHEREY-NAGEL
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Weighing Paper, Grade 531
Supplier: VWR Collection
VWR®, Bottle-Top Vacuum Filtration Systems, PES
Supplier: VWR Collection
VWR®, Glass Fibre Filters
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Vacuum filtration units, Rapid-Flow™ filters MF 75, Nalgene®
Supplier: Thermo Fisher Scientific
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Syringe Filters
Supplier: VWR Collection
Glass microfibre filters without binder, grade GF/C, Whatman™
Supplier: Whatman products (Cytiva)
Hydrochloric acid 32%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Accessories for MA series
Supplier: Sartorius Balances
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Solva™, Avantor®
Supplier: Avantor Fluid Handling