Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

80 mm, 20 mm, 210 mm
Catalog # 221-1141
Supplier:   SEMADENI
Funnels with short stem
Catalog # 221-1141
Supplier:  SEMADENI
Funnels with short stem
Funnels with short stem

80 mm, 20 mm, 210 mm

Price
£13.50
Log in for availability
1 * 1 items
Product Details & Documents

Funnels, made of HDPE, with smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.

  • Easy to clean
  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Funnel O-Ø:
    210 mm
  • Stem O-Ø:
    20 mm
  • Stem length:
    80 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.