£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
Tin(IV) isopropoxide, 98% (metals basis), 10% w/v in isopropanol/toluene 1 * 25 mL
36563.AC
:
Thermo Fisher Scientific
Tin(IV) isopropoxide, 98% (metals basis), 10% w/v in isopropanol/toluene 1 * 25 mL
36563.AC
:
Thermo Fisher Scientific
msds_ALFA036563
Frequently Bought Together
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
: Merck
Dimethyl sulfoxide EMPLURA 1 * 185 L
: Merck
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
: Thermo Fisher Scientific
Tin(IV) isopropoxide isopropanol adduct ≥98% (metals basis)
: Thermo Fisher Scientific
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
: Honeywell Chemicals
Toluene, low water content ≥99.5% (by GC, corrected for water content), BakerDRY™, J.T.Baker®
: Avantor
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Toluene, anhydrous ≥99.8%, over molecular sieves
: Thermo Fisher Scientific