Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Bismuth(III) carbonate basic ≥98.5%

Bismuth(III) carbonate basic ≥98.5%

Glass bottle for solids, 500 g
Catalog # 33216.36
Bismuth(III) carbonate basic ≥98.5%
Catalog # 33216.36
CAS Number:  5892-10-4
Bismuth(III) carbonate basic ≥98.5%
Bismuth(III) carbonate basic ≥98.5%

Glass bottle for solids, 500 g

CAS Number:  5892-10-4
Price
£263.00
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98.5% min CAS No.: 5892-10-4 Molecular Formula: (BiO)2CO3 (approximate) Formula Weight: 509.97 Physical Form: Powder Density: 6.86 Solubility: Practically insoluble in water, alcohol. Soluble in nitric acid, hydrochloric acid, concentrated acetic acid Merck Reference: 14,1282 MDL No.: MFCD00010877

Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazes

Synonyms:  Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate
  • CAS Number:5892-10-4
  • Formula:(BiO)₂CO₃
  • MDL Number:MFCD00010877
  • Molecular Weight:509.97 g/mol
  • Merck Index:14,1282
  • Density:6,86
  • Storage Temperature:Ambient
  • EINECS:227-567-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.