Bismuth(III) carbonate basic ≥98.5%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Bismuth(III) carbonate basic ≥98.5%
Catalog # 33216.22
Supplier: Thermo Fisher Scientific
CAS Number: 5892-10-4
Jump to:
Product Details & Documents
Purity: 98.5% min CAS No.: 5892-10-4 Molecular Formula: (BiO)2CO3 (approximate) Formula Weight: 509.97 Physical Form: Powder Density: 6.86 Solubility: Practically insoluble in water, alcohol. Soluble in nitric acid, hydrochloric acid, concentrated acetic acid Merck Reference: 14,1282 MDL No.: MFCD00010877
Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazes
Synonyms: Bismuth carbonate oxide, Dibismuth carbonate dioxide, Bismuth subcarbonate
- CAS Number:5892-10-4
- Formula:(BiO)₂CO₃
- MDL Number:MFCD00010877
- Molecular Weight:509.97 g/mol
- Merck Index:14,1282
- Density:6,86
- Storage Temperature:Ambient
- EINECS:227-567-9
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



