Orders LinkContactUsLinkComponent
Rods, ceramic, round, solid, AL23
6,0 mm, 800 mm
  32933.EM
Rods, ceramic, round, solid, AL23
  32933.EM
Rods, ceramic, round, solid, AL23
6,0 mm, 800 mm
1 * 800 mm
Pure Al₂O₃ Aluminium oxide, AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
  • Ext.Ø:
    6,0 mm
  • Length:
    800 mm
  • Seq:
    53