ContactUsLinkComponent
Spectroflux 108, Lithium tetraborate, Lanthanum oxide and Sodium nitrate, 76.2:14.3:9.5 w/w% 1 * 1 kg
Catalog # 12085.A1
Spectroflux 108, Lithium tetraborate, Lanthanum oxide and Sodium nitrate, 76.2:14.3:9.5 w/w% 1 * 1 kg
Catalog # 12085.A1