ContactUsLinkComponent
Spectroflux 100B, Lithium metaborate and Lithium tetraborate, 80:20 w/w% 1 * 5 items
Catalog # 12080.D9
Spectroflux 100B, Lithium metaborate and Lithium tetraborate, 80:20 w/w% 1 * 5 items
Catalog # 12080.D9