ContactUsLinkComponent
Pipette, Research Variable 3111 Volume 2 - 20 ul - Eppendorf 1 * 1 items
Catalog # 613-3584
Supplier:  EPPENDORF
Pipette, Research Variable 3111 Volume 2 - 20 ul - Eppendorf 1 * 1 items
Catalog # 613-3584
Supplier:  EPPENDORF
Frequently Bought Together