ContactUsLinkComponent
Single burette holder 3-pointgrip rubber covered jaws 1 * 1 items
Catalog # 3598..
Supplier:  R L ENTERPRISES
Single burette holder 3-pointgrip rubber covered jaws 1 * 1 items
Catalog # 3598..
Supplier:  R L ENTERPRISES
Frequently Bought Together