Vial-Crimp Type 2.0 Ml Pkg/144 (PERKIN N9301385) 1 * 100 items
Catalog # PERCN9301385
Supplier: PerkinElmer
Vial-Crimp Type 2.0 Ml Pkg/144 (PERKIN N9301385) 1 * 100 items
Catalog # PERCN9301385
Supplier: PerkinElmer
Frequently Bought Together
Measuring cylinders, tall form, class B, PP
Supplier: Brand
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Safety spectacles, Virtua™ AP
Supplier: 3M
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Disposable cuvettes
Supplier: RATIOLAB
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Supplier: VWR Collection
MAKE-UP 1 * 750 mL
Supplier: MARKET SOURCE PART PROCESS
Volumetric flasks acc. to Kohlrausch
Supplier: witeg Labortechnik
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Autosampler caps and septa
Supplier: PerkinElmer
Titrants and reagents for volumetric Karl Fischer titrations, HYDRANAL™, Fluka™
Supplier: Honeywell Chemicals
Snap caps with 11 mm septa
Supplier: Agilent